4-Bromothiazole 98%

Supplier: Ambeed
Warning

A172160-100G A172160-10G A172160-5G A172160-1G A172160-25G
103806-702EA 12 USD
103806-702 103844-986 76974-068 76974-070 77082-170 77301-188 77301-190
4-Bromothiazole 98%
4-Bromothiazole

Precaución: for Research Use Only

Formula: C₃H₂BrNS
Boiling Pt: 197.4 °C (760 mm Hg)
Flash Pt: 101.6 °C
Storage Temperature: Ambient
MDL Number: MFCD06657592
CAS Number: 34259-99-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR