4-Bromotetrahydropyran 97%

Supplier: Ambeed
Warning

Synonyms: 4-Bromooxane, 4-Bromotetrahydro-2H-pyran

A548790-1G A548790-5G A548790-100G A548790-10G A548790-25G
103828-680EA 11 USD
103828-680 103839-802 76843-876 76843-878 77169-704 77169-706 77169-708
4-Bromotetrahydropyran 97%
4-Bromotetrahydropyran

Precaución: for Research Use Only

Formula: C₅H₉BrO
MW: 165.0300 g/mol
Boiling Pt: 178 °C (760 mm Hg)
Flash Pt: 71.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07371501
CAS Number: 25637-16-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR