4-Bromoquinoline-6-carboxylic acid 95%

Supplier: Ambeed
Warning

A209463-250MG A209463-5G A209463-1G
103817-942EA 93 USD
103817-942 76785-644 76785-646 77303-098
4-Bromoquinoline-6-carboxylic acid 95%
4-Bromoquinoline-6-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₆BrNO₂
MW: 252.0600 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11518955
CAS Number: 219763-87-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR