4-Bromoquinoline 95%

Supplier: Ambeed
Warning

A459595-1G A459595-25G A459595-5G A459595-100G A459595-10G
103827-902EA 6 USD
103827-902 103838-978 76842-848 76842-850 76842-852 77291-770 77291-772
4-Bromoquinoline 95%
4-Bromoquinoline

Precaución: for Research Use Only

Formula: C₉H₆BrN
MW: 208.06 g/mol
Boiling Pt: 295.9 °C (760 mm Hg)
Melting Pt: 29…34 °C
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07644514
CAS Number: 3964-04-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR