4-Bromoquinolin-2-one 95%

Supplier: Ambeed
Warning

A111821-100MG A111821-1G A111821-250MG A111821-5G
103797-906EA 175 USD
103797-906 103823-728 77284-184 77284-186 77284-188 77284-190
4-Bromoquinolin-2-one 95%
4-Bromoquinolin-2-one

Precaución: for Research Use Only

Formula: C₉H₆BrNO
Boiling Pt: 381.2 °C (760 mm Hg)
Flash Pt: 184.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08234664
CAS Number: 938-39-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR