4-Bromoquinazoline 97%

Supplier: Ambeed
Warning

A148856-250MG A148856-100MG
103803-974EA 87 USD
103803-974 103842-094 77748-037 77748-036
4-Bromoquinazoline 97%
4-Bromoquinazoline

Precaución: Inert atmosphere, 2…8 °C.

Formula: C₈H₅BrN₂
Boiling Pt: 304 °C (760 mm Hg)
Flash Pt: 138 °C
Storage Temperature: Refrigerator
MDL Number: MFCD10697879
CAS Number: 354574-59-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR