2-Amino-4-bromopyrimidine 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Bromopyrimidin-2-amine
A161385-100MG
A161385-250MG
103805-480EA
29
USD
103805-480
103843-718
77273-314
77273-316
2-Amino-4-bromopyrimidine 97%
2-Amino-4-bromopyrimidine
Precaución: for Research Use Only
Formula:
C₄H₄BrN₃ MW: 174.00 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD09999168 CAS Number: 343926-69-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...