4-Bromopyridinium chloride 95%
Supplier: Ambeed
|
Warning
|
A156741-100G
A156741-10G
A156741-500G
A156741-25G
103843-120EA
5
USD
103843-120
77028-406
77028-408
77028-410
77318-918
4-Bromopyridinium chloride 95%
4-Bromopyridinium chloride
Precaución: for Research Use Only
Formula:
C₅H₅BrClN Melting Pt: 270 °C Flash Pt: 65.8 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012828 CAS Number: 19524-06-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...