4-Bromopyridin-2(1H)-one 96%
Supplier: Ambeed
|
Warning
|
A225850-250MG
A225850-1G
103819-206EA
7
USD
103819-206
103850-108
76786-126
77088-826
4-Bromopyridin-2(1H)-one 96%
4-Bromopyridin-2(1H)-one
Precaución: for Research Use Only
Formula:
C₅H₄BrNO MW: 174 g/mol Boiling Pt: 305.9 °C (760 mm Hg) Melting Pt: 186 °C Flash Pt: 138.8 °C Storage Temperature: Ambient |
MDL Number:
MFCD11226860 CAS Number: 36953-37-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...