4-Bromo-p-terphenyl 98%

Supplier: Ambeed
Warning

A295103-1G A295103-5G A295103-100G A295103-10G A295103-250MG
103814-044EA 6 USD
103814-044 103835-180 76837-832 76837-834 77289-856 77289-858 77289-860
4-Bromo-p-terphenyl 98%
4-Bromo-p-terphenyl

Precaución: for Research Use Only

Formula: C₁₈H₁₃Br
MW: 309.1998 g/mol
Boiling Pt: 441.9 °C (760 mm Hg)
Melting Pt: 232 °C
Flash Pt: 217.7 °C
Storage Temperature: Ambient
MDL Number: MFCD02685117
CAS Number: 1762-84-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR