4-Bromopiperidine hydrobromide 97%

Supplier: Ambeed
Warning

A138145-100G A138145-10G A138145-500G A138145-25G A138145-5G
103802-566EA 5 USD
103802-566 77162-208 77162-210 77162-212 77298-966 77315-588
4-Bromopiperidine hydrobromide 97%
4-Bromopiperidine hydrobromide

Precaución: for Research Use Only

Formula: C₅H₁₁Br₂N
MW: 244.9600 g/mol
Melting Pt: 199…202 °C
Flash Pt: 64.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00191858
CAS Number: 54288-70-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR