4-Bromopicolinic acid 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Bromopyridine-2-carboxylic acid
A113480-100G
A113480-10G
A113480-25G
A113480-500G
A113480-1G
A113480-5G
103798-240EA
5
USD
103798-240
103824-082
76995-360
76995-362
76995-364
76995-366
77310-898
77310-900
4-Bromopicolinic acid 95%
4-Bromopicolinic acid
Precaución: for Research Use Only
Formula:
C₆H₄BrNO₂ Boiling Pt: 347.8 °C (760 mm Hg) Melting Pt: 172…174 °C Storage Temperature: Ambient |
MDL Number:
MFCD03426665 CAS Number: 30766-03-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...