4-Bromophthalonitrile 96%

Supplier: Ambeed
Warning

A146891-100MG A146891-250MG A146891-1G A146891-25G A146891-5G
103803-714EA 15 USD
103803-714 103841-808 77285-974 77285-976 77317-158 77317-160 77317-162
4-Bromophthalonitrile 96%
4-Bromophthalonitrile

Precaución: for Research Use Only

Formula: C₈H₃BrN₂
Flash Pt: 150.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00221794
CAS Number: 70484-01-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR