(4-Bromophenyl)acetic acid 98%
Supplier: Ambeed
|
Warning
|
A259236-25G
A259236-100G
A259236-500G
A259236-10G
103809-838EA
147
USD
103809-838
77091-846
77238-464
77238-466
77550-634
(4-Bromophenyl)acetic acid 98%
(4-Bromophenyl)acetic acid
Precaución: for Research Use Only
Formula:
(C₆H₄Br)CH₂COOH Boiling Pt: 326 °C (760 mm Hg) Melting Pt: 114…117 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004342 CAS Number: 1878-68-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...