4-Bromo-N-methylpyridin-2-amine 97%

Supplier: Ambeed
Warning

A278985-250MG A278985-1G
103813-444EA 108 USD
103813-444 103853-388 77014-562 77305-312
4-Bromo-N-methylpyridin-2-amine 97%
4-Bromo-N-methylpyridin-2-amine

Precaución: for Research Use Only

Formula: C₆H₇BrN₂
MW: 187.037 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09834845
CAS Number: 468718-65-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR