4-Bromo-N-methylbenzenesulfonamide 98%

Supplier: Ambeed
Warning

A446319-1G A446319-25G A446319-5G
103827-742EA 10 USD
103827-742 103838-808 76842-602 76842-604 76842-606
4-Bromo-N-methylbenzenesulfonamide 98%
4-Bromo-N-methylbenzenesulphonamide

Precaución: for Research Use Only

Formula: C₇H₈BrNO₂S
Boiling Pt: 330.3 °C (760 mm Hg)
Melting Pt: 70…72 °C (760 mm Hg)
Flash Pt: 153.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00993162
CAS Number: 703-12-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR