4-Bromo-1-naphthol 97%

Supplier: Ambeed
Warning

Synonyms: 4-Bromonaphthalen-1-ol

A292847-1G A292847-5G A292847-100G A292847-10G A292847-250MG
103813-958EA 6 USD
103813-958 103835-098 76837-702 76837-704 77114-280 77114-282 77114-284
4-Bromo-1-naphthol 97%
4-Bromo-1-naphthol

Precaución: for Research Use Only

Formula: C₁₀H₇BrO
MW: 223.0700 g/mol
Melting Pt: 128…133 °C
Flash Pt: 125 °C
Storage Temperature: Ambient
MDL Number: MFCD03129265
CAS Number: 571-57-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR