4-Bromo-N,N-dimethylbenzamide 98%

Supplier: Ambeed
Warning

A666655-1G A666655-5G A666655-10G A666655-250MG A666655-25G
103840-476EA 130 USD
103840-476 76844-682 76844-684 77035-574 77035-576 77035-578
4-Bromo-N,N-dimethylbenzamide 98%
4-Bromo-N,N-dimethylbenzamide

Precaución: for Research Use Only

Formula: C₉H₁₀BrNO
MW: 228.0900 g/mol
Boiling Pt: 320.9 °C (760 mm Hg)
Flash Pt: 147.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00463696
CAS Number: 18469-37-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR