4-Bromoisoquinoline 97%

Supplier: Ambeed
Warning

A104685-100G A104685-10G A104685-25G A104685-500G A104685-5G
103822-482EA 7 USD
103822-482 76687-184 76687-186 76687-188 76687-190 77309-346
4-Bromoisoquinoline 97%
4-Bromoisoquinoline

Precaución: for Research Use Only

Formula: C₉H₆BrN
MW: 208.06 g/mol
Boiling Pt: 282.5 °C (760 mm Hg)
Melting Pt: 40…43 °C
Flash Pt: 113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006904
CAS Number: 1532-97-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR