1-Amino-4-bromoisoquinoline 97%

Supplier: Ambeed
Warning

A152719-10G A152719-250MG A152719-5G A152719-1G A152719-25G
103804-424EA 9 USD
103804-424 103842-596 76997-400 76997-402 77299-974 77318-226 77318-228
1-Amino-4-bromoisoquinoline 97%
1-Amino-4-bromoisoquinoline

Precaución: for Research Use Only

Formula: C₉H₇BrN₂
Boiling Pt: 343.9 °C (760 mm Hg)
Melting Pt: 140…144 °C
Storage Temperature: Ambient
MDL Number: MFCD05155261
CAS Number: 55270-27-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR