4-Bromoisoindolin-1-one 97%

Supplier: Ambeed
Warning

A862601-1G A862601-25G A862601-5G A862601-100G A862601-10G A862601-250MG
103841-258EA 35 USD
103841-258 76845-516 76845-518 76845-520 77294-034 77294-036 77294-038
4-Bromoisoindolin-1-one 97%
4-Bromoisoindolin-1-one

Precaución: for Research Use Only

Formula: C₈H₆BrNO
MW: 212.0400 g/mol
Boiling Pt: 449 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD09701286
CAS Number: 337536-15-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR