4-Bromoindoline 97%

Supplier: Ambeed
Warning

A393566-1G A393566-5G A393566-10G A393566-250MG
103826-924EA 15 USD
103826-924 103837-962 76841-366 76841-368 77168-250 77168-252
4-Bromoindoline 97%
4-Bromoindoline

Precaución: for Research Use Only

Formula: C₈H₈BrN
MW: 198.06 g/mol
Boiling Pt: 278.8 °C (760 mm Hg)
Flash Pt: 122.442 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07371633
CAS Number: 86626-38-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR