4-Bromoindole-3-carboxyaldehyde 98%
Supplier: Ambeed
|
Warning
|
A196612-10G
A196612-250MG
A196612-5G
A196612-1G
103816-858EA
19
USD
103816-858
103847-472
76825-424
76825-426
77085-436
77302-522
4-Bromoindole-3-carboxyaldehyde 98%
4-Bromoindole-3-carboxyaldehyde
Precaución: for Research Use Only
Formula:
C₉H₆BrNO MW: 224.06 g/mol Boiling Pt: 395.6 °C (760 mm Hg) Melting Pt: 179…180 °C Flash Pt: 193 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD05864695 CAS Number: 98600-34-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...