4-Bromo-2-furaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: 4-Bromofurfural, 4-Bromofuran-2-carbaldehyde

A113071-10G A113071-250MG A113071-25G A113071-1G A113071-5G
103798-158EA 39 USD
103798-158 103823-998 77284-278 77284-280 77284-282 77310-830 77310-832
4-Bromo-2-furaldehyde 98%
4-Bromo-2-furaldehyde

Precaución: for Research Use Only

Formula: C₅H₃BrO₂
MW: 174.98 g/mol
Boiling Pt: 225 °C (760 mm Hg)
Melting Pt: 54…58 °C
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00671504
CAS Number: 21921-76-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR