4-Bromobutan-1-amine hydrobromide 96%

Supplier: Ambeed
Warning

A230654-10G A230654-250MG A230654-1G A230654-25G A230654-5G A230654-100G
103819-562EA 16 USD
103819-562 103850-462 76976-144 76976-146 77089-326 77089-328 77089-330 77672-048
4-Bromobutan-1-amine hydrobromide 96%
4-Bromobutan-1-amine hydrobromide

Precaución: for Research Use Only

Formula: C₄H₁₁Br₂N
MW: 232.9448 g/mol
Boiling Pt: 175.8 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD07780174
CAS Number: 24566-81-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR