4-Bromobenzo[b]thiophene-2-carboxylic acid 98%
Supplier: Ambeed
|
Warning
|
A166212-1G
A166212-5G
103806-026EA
14
USD
103806-026
103844-286
77320-528
77320-530
4-Bromobenzo[b]thiophene-2-carboxylic acid 98%
4-Bromobenzo[b]thiophene-2-carboxylic acid
Precaución: for Research Use Only
Formula:
C₉H₅BrO₂S MW: 257.1038 g/mol Boiling Pt: 429.7 °C (760 mm Hg) Melting Pt: 265 °C Flash Pt: 213.7 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD03407342 CAS Number: 5194-37-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...