4-Bromobenzo[b]thiophene 98%

Supplier: Ambeed
Warning

A236336-25G A236336-5G A236336-100G A236336-10G A236336-500G A236336-1G
103811-082EA 6 USD
103811-082 103850-854 77089-902 77089-904 77288-588 77288-590 77288-592 77304-040
4-Bromobenzo[b]thiophene 98%
4-Bromobenzo[b]thiophene

Precaución: for Research Use Only

Formula: C₈H₅BrS
MW: 213.0900 g/mol
Boiling Pt: 284.7 °C (760 mm Hg)
Flash Pt: 126 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08669544
CAS Number: 5118-13-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR