4-Bromoresorcinol 96%
Supplier: Ambeed
|
Warning
|
A101409-100G
A101409-10G
A101409-25G
A101409-1G
A101409-5G
103795-960EA
14
USD
103795-960
103821-644
77282-720
77282-722
77282-724
77297-092
77297-094
4-Bromoresorcinol 96%
4-Bromoresorcinol
Precaución: for Research Use Only
Formula:
C₆H₅BrO₂ Boiling Pt: 150 °C (760 mm Hg) Melting Pt: 97…100 °C Flash Pt: 136.8 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002272 CAS Number: 6626-15-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...