4-Bromo-o-phenylenediamine 98%

Supplier: Ambeed
Danger

Synonyms: 1-Bromo-3,4-diaminobenzene, 4-Bromobenzene-1,2-diamine, 4-Bromo-1,2-phenylenediamine, 4-Bromo-1,2-diaminobenzene, 2-Amino-4-bromophenylamine

A141113-10G A141113-1G A141113-25G A141113-5G
77109-812EA 8.24 USD
77109-812 77109-814 77316-086 77316-088
4-Bromo-o-phenylenediamine 98%
4-Bromo-o-phenylenediamine

Precaución: for Research Use Only

Formula: C₆H₇BrN₂
Boiling Pt: 289.3 °C (760 mm Hg)
Melting Pt: 65…69 °C
Storage Temperature: Ambient
MDL Number: MFCD02660622
CAS Number: 1575-37-7
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR