4-Bromobenzaldehyde 98%

Supplier: Ambeed
Warning

A128214-100G A128214-500G A128214-1000G A128214-25G
103808-368EA 141 USD
103808-368 77149-476 77149-478 77298-272 77313-380
4-Bromobenzaldehyde 98%
4-Bromobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₅BrO
Melting Pt: 55…58 °C
Flash Pt: 109 °C
Storage Temperature: Ambient
MDL Number: MFCD00003377
CAS Number: 1122-91-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR