4-Bromo-7-methylindole 95%

Supplier: Ambeed
Warning

Synonyms: 4-Bromo-7-methyl-1H-indole

A111712-100MG A111712-10G A111712-1G A111712-250MG
103797-890EA 87 USD
103797-890 103823-708 77284-162 77284-164 77284-166 77284-168
4-Bromo-7-methylindole 95%
4-Bromo-7-methylindole

Precaución: for Research Use Only

Formula: C₉H₈BrN
MW: 210.071 g/mol
Boiling Pt: 325 °C (760 mm Hg)
Flash Pt: 150.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08272256
CAS Number: 936092-87-4

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR