4-Bromo-7-methoxy-1H-pyrrolo[2,3-c]pyridine 98%
Supplier: Ambeed
|
Warning
|
A140757-100MG
A140757-250MG
A140757-1G
A140757-5G
103802-900EA
81
USD
103802-900
76996-856
76996-858
77316-010
77316-012
4-Bromo-7-methoxy-1H-pyrrolo[2,3-c]pyridine 98%
4-Bromo-7-methoxy-1H-pyrrolo[2,3-c]pyridine
Precaución: for Research Use Only
Formula:
C₈H₇BrN₂O Melting Pt: 174…175 °C Storage Temperature: Ambient |
MDL Number:
MFCD11845544 CAS Number: 425380-37-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...