4-Bromo-7H-pyrrolo[2,3-d]pyrimidine 96%

Supplier: Ambeed
Warning

A162566-1G A162566-5G
103805-630EA 24 USD
103805-630 103843-880 77319-930 77319-932
4-Bromo-7H-pyrrolo[2,3-d]pyrimidine 96%
4-Bromo-7H-pyrrolo[2,3-d]pyrimidine

Precaución: for Research Use Only

Formula: C₆H₄BrN₃
MW: 198.0200 g/mol
Melting Pt: 167 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08445698
CAS Number: 889939-42-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR