4-Bromo-7-fluoroindole 98%

Supplier: Ambeed
Warning

Synonyms: 4-Bromo-7-fluoro-1H-indole

A142516-250MG A142516-50MG A142516-1G A142516-5G
103803-174EA 103 USD
103803-174 77027-632 77027-634 77316-400 77316-402
4-Bromo-7-fluoroindole 98%
4-Bromo-7-fluoroindole

Precaución: for Research Use Only

Formula: C₈H₅BrFN
MW: 214.04 g/mol
Boiling Pt: 314.2 °C (760 mm Hg)
Melting Pt: 24…25 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03095318
CAS Number: 883500-66-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR