4-Bromo-7-chloroquinoline 95%

Supplier: Ambeed
Warning

A205016-10G A205016-1G A205016-25G A205016-5G
103817-584EA 20 USD
103817-584 103848-248 77112-294 77302-926 77302-928 77302-930
4-Bromo-7-chloroquinoline 95%
4-Bromo-7-chloroquinoline

Precaución: for Research Use Only

Formula: C₉H₅BrClN
Melting Pt: 114…115 °C
Storage Temperature: Ambient
MDL Number: MFCD07700232
CAS Number: 98519-65-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR