4-Bromo-7-chloro-1H-pyrrolo[2,3-c]pyridine 98%

Supplier: Ambeed
Warning

A127806-100MG A127806-250MG A127806-5G A127806-1G
103801-014EA 146 USD
103801-014 76996-188 76996-190 77298-234 77313-280
4-Bromo-7-chloro-1H-pyrrolo[2,3-c]pyridine 98%
4-Bromo-7-chloro-1H-pyrrolo[2,3-c]pyridine

Precaución: for Research Use Only

Formula: C₇H₄BrClN₂
Storage Temperature: Refrigerator
MDL Number: MFCD11848529
CAS Number: 425380-38-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR