4-Bromo-7-chloro-1H-indazole 95%

Supplier: Ambeed
Warning

A227299-250MG A227299-100MG
103819-312EA 63 USD
103819-312 103850-218 77013-418 77892-368
4-Bromo-7-chloro-1H-indazole 95%
4-Bromo-7-chloro-1H-indazole

Precaución: for Research Use Only

Formula: C₇H₄BrClN₂
Storage Temperature: Ambient
MDL Number: MFCD16658654
CAS Number: 1186334-61-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR