4-Bromo-7-azaindole 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Bromo-7-azaindole, 4-Bromo-1H-pyrrolo[2,3-b]pyridine
A287737-1G
A287737-5G
A287737-100G
A287737-10G
A287737-250MG
A287737-25G
A287737-500G
103813-774EA
10
USD
103813-774
103834-916
76837-464
76837-466
77166-734
77166-736
77166-738
77166-740
77166-742
4-Bromo-7-azaindole 98%
4-Bromo-7-azaindole
Precaución: for Research Use Only
Formula:
C₇H₅BrN₂ Melting Pt: 178…183 °C Storage Temperature: Ambient |
MDL Number:
MFCD08272233 CAS Number: 348640-06-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...