4-Bromo-6-methylpyridin-2-ol 95%
Supplier: Ambeed
|
Warning
|
A178291-10G
A178291-250MG
A178291-1G
A178291-5G
A178291-25G
103807-314EA
26
USD
103807-314
103845-654
76813-452
76813-454
77083-064
77083-066
77883-354
4-Bromo-6-methylpyridin-2-ol 95%
4-Bromo-6-methylpyridin-2-ol
Precaución: for Research Use Only
Formula:
C₆H₆BrNO Storage Temperature: Refrigerator |
MDL Number:
MFCD16610030 CAS Number: 865156-59-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...