4-Bromo-6-methylpyridin-2-amine 97%

Supplier: Ambeed
Warning

A126418-100MG A126418-250MG A126418-5G A126418-10G
103800-810EA 110 USD
103800-810 76810-934 76810-936 77313-022 77566-448
4-Bromo-6-methylpyridin-2-amine 97%
4-Bromo-6-methylpyridin-2-amine

Precaución: For research use only.

Formula: C₆H₇BrN₂
MW: 187.037 g/mol
Boiling Pt: 268 °C (760 mm Hg)
Flash Pt: 116 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08062948
CAS Number: 524718-27-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR