4-Bromo-6-methoxypyridin-2-amine 97%

Supplier: Ambeed
Warning

A552336-1G A552336-100MG A552336-250MG A552336-50MG
103828-696EA 253 USD
103828-696 76843-904 77144-498 77144-500 77144-502
4-Bromo-6-methoxypyridin-2-amine 97%
4-Bromo-6-methoxypyridin-2-amine

Precaución: for Research Use Only

Formula: C₆H₇BrN₂O
MW: 203.04 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18258916
CAS Number: 1211526-95-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR