4-Bromo-6-fluoro-1H-indole 95%

Supplier: Ambeed
Warning

A186894-10G A186894-250MG A186894-1G A186894-25G A186894-5G
103815-972EA 58 USD
103815-972 103846-560 76825-114 76825-116 77302-046 77302-048 77302-050
4-Bromo-6-fluoro-1H-indole 95%
4-Bromo-6-fluoro-1H-indole

Precaución: for Research Use Only

Formula: C₈H₅BrFN
MW: 214.0344 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08272203
CAS Number: 885520-70-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR