4-Bromo-6-chloropyridazin-3(2H)-one 97%

Supplier: Ambeed
Warning

A301258-100MG A301258-10G A301258-250MG A301258-1G A301258-25G A301258-5G
103814-300EA 10 USD
103814-300 103835-450 76816-778 76816-780 76816-782 76838-130 76838-132 76838-134
4-Bromo-6-chloropyridazin-3(2H)-one 97%
4-Bromo-6-chloropyridazin-3(2H)-one

Precaución: for Research Use Only

Formula: C₄H₂BrClN₂O
Storage Temperature: Refrigerator
MDL Number: MFCD21364604
CAS Number: 933041-13-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR