4-Bromo-6-chloronicotinic acid 95%
Supplier: Ambeed
|
Warning
|
A251445-1G
A251445-100MG
A251445-250MG
A251445-5G
103811-982EA
228
USD
103811-982
77091-208
77165-982
77165-984
77165-986
4-Bromo-6-chloronicotinic acid 95%
4-Bromo-6-chloronicotinic acid
Precaución: for Research Use Only
Formula:
C₆H₃BrClNO₂ MW: 236.45 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD18257866 CAS Number: 1256834-13-5 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...