4-Bromo-6-chloronicotinaldehyde 98%

Supplier: Ambeed
Warning

A102092-100MG A102092-250MG
103796-294EA 81 USD
103796-294 76982-212 77884-926
4-Bromo-6-chloronicotinaldehyde 98%
4-Bromo-6-chloronicotinaldehyde

Precaución: for Research Use Only

Formula: C₆H₃BrClNO
Storage Temperature: Refrigerator
MDL Number: MFCD13189209
CAS Number: 1060805-64-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR