4-Bromo-6-chloro-1H-indazole 98%

Supplier: Ambeed
Warning

A146808-100G A146808-10G A146808-250MG A146808-1G A146808-25G A146808-5G
103803-702EA 52 USD
103803-702 103841-796 77285-954 77285-956 77285-958 77317-130 77317-132 77317-134
4-Bromo-6-chloro-1H-indazole 98%
4-Bromo-6-chloro-1H-indazole

Precaución: for Research Use Only

Formula: C₇H₄BrClN₂
MW: 231.4800 g/mol
Melting Pt: 219…221 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07781352
CAS Number: 885519-03-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR