4-Bromo-6,7-dimethoxyquinoline 97%

Supplier: Ambeed
Warning

A205141-10G A205141-250MG A205141-1G A205141-25G
103817-602EA 87 USD
103817-602 103848-266 76800-496 76800-498 77086-466 77302-940
4-Bromo-6,7-dimethoxyquinoline 97%
4-Bromo-6,7-dimethoxyquinoline

Precaución: for Research Use Only

Formula: C₁₁H₁₀BrNO₂
MW: 268.1100 g/mol
Boiling Pt: 359.2 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD08063223
CAS Number: 666734-51-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR