4-Bromo-6-(trifluoromethyl)-1H-indole 98%

Supplier: Ambeed
Warning

A101212-100MG A101212-250MG A101212-5G
103795-858EA 55 USD
103795-858 103821-542 76982-172 76982-174 77282-680
4-Bromo-6-(trifluoromethyl)-1H-indole 98%
4-Bromo-6-(trifluoromethyl)-1H-indole

Precaución: for Research Use Only

Formula: C₉H₅BrF₃N
MW: 264.04 g/mol
Boiling Pt: 298.1 °C (760 mm Hg)
Flash Pt: 134.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09026955
CAS Number: 1000342-93-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR