4-Bromo-6-(trifluoromethyl)-1H-indazole 97%

Supplier: Ambeed
Warning

Synonyms: 4-Bromo-6-trifluoromethyl-1H-indazole

A417797-5G A417797-1G A417797-100MG
103827-344EA 46 USD
103827-344 103838-390 76841-986 77307-194 77673-026
4-Bromo-6-(trifluoromethyl)-1H-indazole 97%
4-Bromo-6-(trifluoromethyl)-1H-indazole

Precaución: for Research Use Only

Formula: C₈H₄BrF₃N₂
MW: 265.03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09026956
CAS Number: 1000342-95-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR