4-Bromo-5-methylbenzene-1,2-diamine 95%

Supplier: Ambeed
Warning

A303200-1G A303200-25G A303200-5G A303200-10G A303200-250MG
103814-364EA 8 USD
103814-364 103835-528 76838-210 76838-212 76838-214 76988-818 76988-820
4-Bromo-5-methylbenzene-1,2-diamine 95%
4-Bromo-5-methylbenzene-1,2-diamine

Precaución: for Research Use Only

Formula: C₇H₉BrN₂
Storage Temperature: Ambient
MDL Number: MFCD16987757
CAS Number: 102169-44-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR